Learn More
MSC Norton Paper Floor Edger Disc, Zirconia Alumina, F-Weight Backing
Catalog No. 502839558
Encompass
When a fast cut is needed on hard wood surfaces, Norton BlueFire Paper Floor Edger Discs are an excellent choice for fast edging on hard woods or concrete. These discs are available in multiple grits, ranging from coarse to medium.
  • Next generation, self-sharpening zirconia alumina abrasive provides a fast cut rate and outlasts conventional discs.
  • Strong F-weight paper backing resists tearing.
  • Open-coat design and anti-static technology minimize loading and dust.
  • Leaves a fine scratch pattern with fewer visible sanding marks for a better finish.
Catalog No. 50-283-9558 Supplier MSC Supplier No. 85150589
When a fast cut is needed on hard wood surfaces, Norton BlueFire Paper Floor Edger Discs are an excellent choice for fast edging on hard woods or concrete. These discs are available in multiple grits, ranging from coarse to medium.
  • Next generation, self-sharpening zirconia alumina abrasive provides a fast cut rate and outlasts conventional discs.
  • Strong F-weight paper backing resists tearing.
  • Open-coat design and anti-static technology minimize loading and dust.
  • Leaves a fine scratch pattern with fewer visible sanding marks for a better finish.