Learn More
MSC Norton Paper Floor Edger Disc, 6-7/8 in, 7/8 in Arbor Hole, 40 Grit, Zirconia Alumina
Catalog No. 502839780
Encompass
This BlueFire Paper Floor Edger Disc is ideal for fast cutting on hard wood surfaces and concrete. It is designed for floor sanding applications using an edger machine.
  • Next-generation, self-sharpening zirconia alumina abrasive provides a fast cut rate and lasts twice as long as conventional zirconia discs.
  • Strong F-weight paper backing resists tearing for extended disc life.
  • Open-coat design and anti-static technology minimize loading and dust, keeping debris away from the floor's surface.
  • Leaves a fine scratch pattern with fewer visible sanding marks for a better finish.
Catalog No. 50-283-9780 Supplier MSC Supplier No. 76600022
This BlueFire Paper Floor Edger Disc is ideal for fast cutting on hard wood surfaces and concrete. It is designed for floor sanding applications using an edger machine.
  • Next-generation, self-sharpening zirconia alumina abrasive provides a fast cut rate and lasts twice as long as conventional zirconia discs.
  • Strong F-weight paper backing resists tearing for extended disc life.
  • Open-coat design and anti-static technology minimize loading and dust, keeping debris away from the floor's surface.
  • Leaves a fine scratch pattern with fewer visible sanding marks for a better finish.