Learn More
MSC Norton Paper Floor Edger Disc, Zirconia Alumina, 100 Grit, 7 in Diameter, 5/16 in Arbor Hole
Catalog No. 502840053
Encompass
This paper floor edger disc is an excellent choice for fast edging on hard woods or concrete surfaces. It features a next-generation, self-sharpening zirconia alumina abrasive that delivers a fast cut rate and extends disc life, leaving a fine scratch pattern for a better finish. The strong F-weight paper backing resists tearing, and the open-coat design with anti-static technology minimizes loading and dust accumulation.
  • Next-generation, self-sharpening zirconia alumina abrasive delivers a fast cut rate and outlasts conventional zirconia discs 2-to-1.
  • Strong F-weight paper backing resists tearing to deliver extended disc life.
  • Open-coat design and anti-static technology work to minimize loading and dust to keep debris away from the floor's surface.
  • Leaves a fine scratch pattern with less visible sanding marks for a better-looking finish.
Catalog No. 50-284-0053 Supplier MSC Supplier No. 85152197
This paper floor edger disc is an excellent choice for fast edging on hard woods or concrete surfaces. It features a next-generation, self-sharpening zirconia alumina abrasive that delivers a fast cut rate and extends disc life, leaving a fine scratch pattern for a better finish. The strong F-weight paper backing resists tearing, and the open-coat design with anti-static technology minimizes loading and dust accumulation.
  • Next-generation, self-sharpening zirconia alumina abrasive delivers a fast cut rate and outlasts conventional zirconia discs 2-to-1.
  • Strong F-weight paper backing resists tearing to deliver extended disc life.
  • Open-coat design and anti-static technology work to minimize loading and dust to keep debris away from the floor's surface.
  • Leaves a fine scratch pattern with less visible sanding marks for a better-looking finish.