Learn More
MSC Norton BlueFire Paper Hook-and-Loop Floor Edger Disc, Zirconia Alumina
Catalog No. 502834810
Encompass
Borton BlueFire Paper Hook-and-Loop Floor Edger Discs are designed for optimal finishing on hard wood or concrete floors. They feature easy hook-and-loop mounting and a durable zirconia alumina abrasive, ensuring efficient and high-quality results with extended disc life.
  • Excellent finish on hard wood or concrete floors.
  • Hook-and-loop backing for easy edger mounting.
  • Self-sharpening zirconia alumina abrasive for fast cutting.
  • 2-to-1 longer life than conventional zirconia discs.
  • Heavy F-weight paper backing resists tearing.
  • Open coat design and anti-static technology minimize loading and dust.
  • Leaves a fine scratch pattern for a better finish.
Catalog No. 50-283-4810 Supplier MSC Supplier No. 98433576
Borton BlueFire Paper Hook-and-Loop Floor Edger Discs are designed for optimal finishing on hard wood or concrete floors. They feature easy hook-and-loop mounting and a durable zirconia alumina abrasive, ensuring efficient and high-quality results with extended disc life.
  • Excellent finish on hard wood or concrete floors.
  • Hook-and-loop backing for easy edger mounting.
  • Self-sharpening zirconia alumina abrasive for fast cutting.
  • 2-to-1 longer life than conventional zirconia discs.
  • Heavy F-weight paper backing resists tearing.
  • Open coat design and anti-static technology minimize loading and dust.
  • Leaves a fine scratch pattern for a better finish.