Learn More
MSC Norton Paper H&L Disc, Zirconia Alumina, 8 in Diameter, 80 Grit
Catalog No. 502838225
Encompass
Norton BlueFire Paper H&L Discs are designed for soft material and wood applications, offering outstanding durability. They are suitable for blending, sanding, and stripping, including on materials like aluminum, fiberglass, and body filler.
  • Next-generation, self-sharpening zirconia alumina abrasive offers 2-3 times the life of aluminum oxide discs with minimal heat
  • Ultra-tough, E-weight latex/paper backing resists tearing while remaining conformable and flexible
  • No risk of color transfer with assured color consistency
  • Hook-and-loop back allows for easy mounting and changing on random orbital sanders or right-angle grinders
Catalog No. 50-283-8225 Supplier MSC Supplier No. 85346278
Norton BlueFire Paper H&L Discs are designed for soft material and wood applications, offering outstanding durability. They are suitable for blending, sanding, and stripping, including on materials like aluminum, fiberglass, and body filler.
  • Next-generation, self-sharpening zirconia alumina abrasive offers 2-3 times the life of aluminum oxide discs with minimal heat
  • Ultra-tough, E-weight latex/paper backing resists tearing while remaining conformable and flexible
  • No risk of color transfer with assured color consistency
  • Hook-and-loop back allows for easy mounting and changing on random orbital sanders or right-angle grinders