Learn More
MSC Norton Hook & Loop Paper Disc, Zirconia Alumina, Heavy Paper E wt.
Catalog No. 502839876
Encompass
These discs are designed for soft material and wood applications, offering outstanding durability for blending, sanding, and stripping. They are also effective on aluminum, fiberglass, and body filler.
  • Next-generation, self-sharpening zirconia alumina abrasive provides 2-3 times the life of aluminum oxide discs with minimal heat.
  • Ultra-tough, E-weight latex/paper backing resists tearing while remaining conformable and flexible.
  • No risk of color transfer with assured color consistency.
  • Hook-and-loop back allows for easy mounting and changing on random orbital sanders or right-angle grinders.
Catalog No. 50-283-9876 Supplier MSC Supplier No. 90086885
These discs are designed for soft material and wood applications, offering outstanding durability for blending, sanding, and stripping. They are also effective on aluminum, fiberglass, and body filler.
  • Next-generation, self-sharpening zirconia alumina abrasive provides 2-3 times the life of aluminum oxide discs with minimal heat.
  • Ultra-tough, E-weight latex/paper backing resists tearing while remaining conformable and flexible.
  • No risk of color transfer with assured color consistency.
  • Hook-and-loop back allows for easy mounting and changing on random orbital sanders or right-angle grinders.