Learn More
MSC Norton Paper H&L Disc, Zirconia Alumina, Self-Sharpening
Catalog No. 502839564
Encompass
BlueFire Paper H&L Discs are designed for soft material and wood applications. These discs provide outstanding durability, making them suitable for blending, sanding, and stripping tasks, even on materials like aluminum, fiberglass, and body filler.
  • Next-generation, self-sharpening zirconia alumina abrasive provides 2-3 times the life of aluminum oxide discs with minimal heat.
  • Ultra-tough, E-weight latex/paper backing resists tearing while staying conformable and flexible.
  • No risk of color transfer with assured color consistency.
  • Hook-and-loop back makes for easy mounting and changing on your random orbital sander or right-angle grinder.
Catalog No. 50-283-9564 Supplier MSC Supplier No. 85346260
BlueFire Paper H&L Discs are designed for soft material and wood applications. These discs provide outstanding durability, making them suitable for blending, sanding, and stripping tasks, even on materials like aluminum, fiberglass, and body filler.
  • Next-generation, self-sharpening zirconia alumina abrasive provides 2-3 times the life of aluminum oxide discs with minimal heat.
  • Ultra-tough, E-weight latex/paper backing resists tearing while staying conformable and flexible.
  • No risk of color transfer with assured color consistency.
  • Hook-and-loop back makes for easy mounting and changing on your random orbital sander or right-angle grinder.