Learn More
Enzo Life Sciences NUCLEAR-ID Blue/Green cell viability reagent (100µl)
SDP
Catalog No. 502012650
Encompass_Preferred
The NUCLEAR-ID  Blue/Green cell viability reagent (ENZ-53004) is a mixture of a blue fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers. Purity: ≥93% (HPLC). Long Term Storage: -80°C. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2650 Supplier Enzo Life Sciences Supplier No. ENZ53004C100
The NUCLEAR-ID  Blue/Green cell viability reagent (ENZ-53004) is a mixture of a blue fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers. Purity: ≥93% (HPLC). Long Term Storage: -80°C. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .