Learn More
Bluecatbio Ma Inc BLUEINTENSE CLEANING SOLUTION
SDP
Catalog No. NC1637033
Encompass
BlueIntense Cleaning solution, 1 bottle. Intensive alkaline microbiocidal and cleaning agent for BlueWasher using the IntenseClean program recommended to be used at least once per week. 500ml bottle makes 5l solution(~ 50 applications)
Catalog No. NC1637033 Supplier Bluecatbio Ma Inc Supplier No. 2BI1
BlueIntense Cleaning solution, 1 bottle. Intensive alkaline microbiocidal and cleaning agent for BlueWasher using the IntenseClean program recommended to be used at least once per week. 500ml bottle makes 5l solution(~ 50 applications)