Learn More
Seracare Life Sciences Inc BLUEPHOS AP ELISA SUBSTRATE
Catalog No. 5067455
Encompass_Preferred
Excellent alternative to pNPP. Designed for use with alkaline phosphatase-labeled conjugates. A soluble, proprietary form of BCIP (5-bromo-4-chloro-3-indoyl phosphate) that features greater sensitivity, wider working range, lower background, and more convenience than pNPP. 2-Component format, simply mix and use.
This item is not returnable. View return policy
Catalog No. 50-674-55 Supplier Seracare Life Sciences Inc Supplier No. 51200060
Excellent alternative to pNPP. Designed for use with alkaline phosphatase-labeled conjugates. A soluble, proprietary form of BCIP (5-bromo-4-chloro-3-indoyl phosphate) that features greater sensitivity, wider working range, lower background, and more convenience than pNPP. 2-Component format, simply mix and use.