Learn More
Sage Science Inc BLUEPIPPIN SIZE SELECTION SYST
SDP
Catalog No. NC1148993
Encompass
  • Automated size selection improves reproducibility and reduces manual effort.
  • Pulsed-field electrophoresis enables resolution of high molecular weight DNA fragments up to 50 kb.
  • Configurable high-pass filtering allows users to collect fragments above a specified size threshold.
  • Enhances sample quality with tight fragment size distributions and minimizes cross-contamination.
  • Each gel cassette processes up to 5 samples without risk of cross-contamination.
  • Integrated system includes built-in CPU, monitor, keyboard, and mouse, eliminating the need for external PC.
Catalog No. NC1148993 Supplier Sage Science Inc Supplier No. BLU0001
Add to Cart
Add to Cart
Request a Quote
  • Automated size selection improves reproducibility and reduces manual effort.
  • Pulsed-field electrophoresis enables resolution of high molecular weight DNA fragments up to 50 kb.
  • Configurable high-pass filtering allows users to collect fragments above a specified size threshold.
  • Enhances sample quality with tight fragment size distributions and minimizes cross-contamination.
  • Each gel cassette processes up to 5 samples without risk of cross-contamination.
  • Integrated system includes built-in CPU, monitor, keyboard, and mouse, eliminating the need for external PC.