Learn More
Crescent Chemical Co Inc BLUESLICK
SDP
Supplier:  Crescent Chemical Co Inc SE42500.01
Encompass_Preferred
BlueSlick Non-toxic BlueSlick™ is the alternative to silane-containing products and is non-irritant to eyes. It can be handled outside of a fume-hood. Suitable to all applications in electrophoresis (DNA sequencing, SDS PAGE, IEF PAGE). It does not affect
Catalog No. 50-248-584