Learn More
Gold Biotechnology Inc BLUEstain™ 2 Protein ladder, 5-245 kDa 500 µL
SDP
Catalog No. 501532820
Encompass_Preferred
The BLUEstainTM 2 Protein ladder is a three-color protein standard with 13 prestained proteins, covering a wide range molecular weights from 3.5 to 245 kDa. Proteins are covalently coupled with a blue chromophore except for two reference bands (one green and one red band at 25 kDa and 75 kDa respectively) when separated on SDS-PAGE (Tris-glycine buffer).
Catalog No. 50-153-2820 Supplier Gold Biotechnology Inc Supplier No. P008500
The BLUEstainTM 2 Protein ladder is a three-color protein standard with 13 prestained proteins, covering a wide range molecular weights from 3.5 to 245 kDa. Proteins are covalently coupled with a blue chromophore except for two reference bands (one green and one red band at 25 kDa and 75 kDa respectively) when separated on SDS-PAGE (Tris-glycine buffer).