Learn More
Gold Biotechnology Inc BLUEstain™ Protein ladder, 11-245 kDa 2x500 µL
SDP
Catalog No. NC1350206
Encompass
The BLUEstainTM Protein ladder is a three-color protein standard with 12 prestained proteins covering a wide range molecular weights from ~11 to 245 kDa. Proteins are covalently coupled with a bluechromophore except for two reference bands (one green and one red band at 25 kDa and 75 kDa respectively) when separated on SDS-PAGE (Tris-glycine buffer). The BLUEstainTM Protein ladder, is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (PVDF, nylon, or nitrocellulose) and for approximating the size of proteins. The ladder is supplied in gel loading buffer and is ready to use.
Catalog No. NC1350206 Supplier Gold Biotechnology Inc Supplier No. P0071000
The BLUEstainTM Protein ladder is a three-color protein standard with 12 prestained proteins covering a wide range molecular weights from ~11 to 245 kDa. Proteins are covalently coupled with a bluechromophore except for two reference bands (one green and one red band at 25 kDa and 75 kDa respectively) when separated on SDS-PAGE (Tris-glycine buffer). The BLUEstainTM Protein ladder, is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (PVDF, nylon, or nitrocellulose) and for approximating the size of proteins. The ladder is supplied in gel loading buffer and is ready to use.