Learn More
Digivac BLUETOOTH BULLSEYE PREC GAUGE
SDP
Catalog No. NC1880953
Encompass_Preferred
Versatile wireless vacuum gauge that you can monitor right from your phone or tablet (Apple ios and android)Enables remote monitoring and troubleshootingSet low and high alarm from your phoneEmail vacuum data to document leaks, baseline pressure or pump-downs.
Catalog No. NC1880953 Supplier Digivac Supplier No. BPGBT
Versatile wireless vacuum gauge that you can monitor right from your phone or tablet (Apple ios and android)Enables remote monitoring and troubleshootingSet low and high alarm from your phoneEmail vacuum data to document leaks, baseline pressure or pump-downs.