Learn More
Bluecatbio Ma Inc BLUEWASHER
SDP
Catalog No. NC1884950
Encompass
Automated, centrifugation-based plate dryer & media changer, software licenses for graphical user interface (GUI) and firmware (FW) with updates for 12 months, gravity based liquid waste handling, dispenser option for 96, 384, and 1536 well plates from up to 7 liquid input channels, standard carrier pair, power and USB cables
Catalog No. NC1884950 Supplier Bluecatbio Ma Inc Supplier No. 1573L
Add to Cart
Add to Cart
Request a Quote
Automated, centrifugation-based plate dryer & media changer, software licenses for graphical user interface (GUI) and firmware (FW) with updates for 12 months, gravity based liquid waste handling, dispenser option for 96, 384, and 1536 well plates from up to 7 liquid input channels, standard carrier pair, power and USB cables