Learn More
Sigma Aldrich Fine Chemicals Biosciences BLUeye Prestained Protein
Catalog No. 502901311
Encompass
The ready-to-use BLUeye prestained protein ladder has 12 sharp bands at 11 17 20 25 35 48 63 75 100 135 180 and 245 kDa which has more bands than commonly used markers. The BLUeye prestained protein ladder is provided in a gel loading buffer. A green and a red reference band respectively enable easier identification. The BLUeye prestained protein ladder was produced by GeneDirex Taiwan.
Catalog No. 50-290-1311 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 94964500UL
The ready-to-use BLUeye prestained protein ladder has 12 sharp bands at 11 17 20 25 35 48 63 75 100 135 180 and 245 kDa which has more bands than commonly used markers. The BLUeye prestained protein ladder is provided in a gel loading buffer. A green and a red reference band respectively enable easier identification. The BLUeye prestained protein ladder was produced by GeneDirex Taiwan.