Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen™ Bluing Reagent

Catalog No. 23730614 Shop All Scigen Inc Products
Change view
Click to view available options
Quantity:
3.8 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-730-614 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-730-614 Supplier Scigen™ Supplier No. 7121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A buffered bluing reagent with a pH of 8 to assure bluing of hematoxylin stained tissue sections.

Scigen™ Bluing Reagent is a buffered bluing reagent with a pH of 8 to assure bluing of hematoxylin stained tissue sections. Suggested bluing time is 30 sounds, after bluing, rinse tissue sections in running water for 1 minute. Bluing reagent must be completely removed from tissue sections prior to Eosin staining. Scigen™ Bluing Reagent is not a concentrate and requires no dilution.

Specifications

Color Clear, Water White
Description Provides Crips Nuclear Detail While Preventing Tissue Separation
For Use With (Application) Enhances the Color of the hematoxylin stained nuclei to a crisp blue-purple
Includes Staining Protocol
Material Aqueous Based
pH 8
Quantity 3.8 L
Solution Type Aqueous
Storage Requirements Room Temperature
Synonym Bluing
Product Type Bluing Reagent
Show More Show Less

FOR IN VITRO LABORATORY USE POISON: NOT FOR DRUG, FOOD, OR HOUSEHOLD USE.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.