Learn More
Scytek Laboratories BLUING REAGENT 1000ML
SDP
Catalog No. NC1872007
Encompass_Preferred
Is a pH-controlled solution for complete, effective bluing of hematoxylin in histological and cytological procedures.
Catalog No. NC1872007 Supplier Scytek Laboratories Supplier No. BRT999
Is a pH-controlled solution for complete, effective bluing of hematoxylin in histological and cytological procedures.