Learn More
Scytek Laboratories BLUING REAGENT 500 ML
SDP
Catalog No. NC1872006
Encompass_Preferred
Is a pH-controlled solution for complete, effective bluing of hematoxylin in histological and cytological procedures.
Catalog No. NC1872006 Supplier Scytek Laboratories Supplier No. BRT500
Is a pH-controlled solution for complete, effective bluing of hematoxylin in histological and cytological procedures.