Learn More
Cayman Chemical Bluma balsamfra Blumatn 10mg
SDP
Catalog No. 502911180
Encompass_Preferred
A flavonoid with antioxidant and anticancer activities; scavenges DPPH radicals in a cell-free assay (SC50 = 90.8 UG/ml); decreases carbon tetrachloride-induced lipid peroxidation in isolated rhesus monkey hepatocytes; cytotoxic to KB oral carcinoma cells (IC50 = 47.72 UG/ml)
Catalog No. 50-291-1180 Supplier Cayman Chemical Supplier No. 3480910 MG
A flavonoid with antioxidant and anticancer activities; scavenges DPPH radicals in a cell-free assay (SC50 = 90.8 UG/ml); decreases carbon tetrachloride-induced lipid peroxidation in isolated rhesus monkey hepatocytes; cytotoxic to KB oral carcinoma cells (IC50 = 47.72 UG/ml)