Learn More
Surgical Design™ Blumenthal Rongeur
Catalog No. 21124461 Shop All Surgical Design Products
Change view
Click to view available options
Blade Style:
Full Curve
Half Curve
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Style
21-124-461 Full Curve
21-124-460 Half Curve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-461 Supplier Surgical Design™ Supplier No. 111219
Add to Cart
Add to Cart
Blumenthal rongeur provides smooth, precise bone cutting with reliable surgical performance.
  • Crafted from medical-grade stainless steel for strength, precision, and corrosion resistance
  • 6-in. length offers excellent control for fine bone removal and trimming
  • Available in Half Curved and Full Curved jaw designs for enhanced surgical access
  • 3 mm bite ideal for controlled, detailed bone resection
  • Sharp, precision-ground jaws ensure smooth and efficient cutting action
  • Ergonomic handle provides comfort and reduces fatigue during extended use
  • Balanced construction for improved handling and accuracy in tight operative fields
  • Common in forensic, pathology, and autopsy bone dissection
  • Ideal for reconstructive, dental, and veterinary bone trimming tasks
  • Suitable for training and research applications requiring precision instrumentation
Specifications
Product Type Bone-cutting Forceps
Blade Material Stainless Steel
Blade Style Full Curve
Length (English) 6 in.
Length (Metric) 15.24 cm
Length (Metric) Blade 3 mm
Material Stainless Steel
Autoclavable Autoclavable