Learn More
Stoelting™ Blumenthal Rongeur
SDP
Catalog No. 10001801 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Slight Angle
Catalog No. Model
10-001-801 Slight Angle
1 options
Catalog No. 10-001-801 Supplier Stoelting™ Supplier No. 5216301
Add to Cart
Add to Cart
Rongeurs feature a sharp-edged, scoop-shaped tip, used for gouging out bone or opening a window in bone, often in the skull. It is used in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.
Specifications
Length (English) 6 in.
Width (Metric) 2 mm
Model Slight Angle