Learn More
International Medical Industries (IMI) Aspirating Needle Blunt Cannula 16 Gauge X 5 Inch (Box of 100)
Catalog No. NC1765111
Encompass
Rx-Tract™ 16 Gauge, 5” (12.7 CM), Blunt Needle provides high-quality solutions for introducing fluids to deep wells and vials, drawing liquids for reconstruction, or pre-fill syringes prior to administration. Rx-Tract Aspirating Needles comes with a conveniently sealed guard to enhance pharmacist and clinician safety. The blunt end prevents accidental needle sticks. •Stainless Steel Needle •Sterile •Non-Pyrogenic •Individual Blister •Box of 100
This item is not returnable. View return policy
Catalog No. NC1765111 Supplier International Medical Industries (IMI) Supplier No. 3231
 Restricted Item
Add to Cart
Add to Cart
Rx-Tract™ 16 Gauge, 5” (12.7 CM), Blunt Needle provides high-quality solutions for introducing fluids to deep wells and vials, drawing liquids for reconstruction, or pre-fill syringes prior to administration. Rx-Tract Aspirating Needles comes with a conveniently sealed guard to enhance pharmacist and clinician safety. The blunt end prevents accidental needle sticks. •Stainless Steel Needle •Sterile •Non-Pyrogenic •Individual Blister •Box of 100