Learn More
STEMCELL Technologies Blunt-End Needles, Size: 2000 Units
SDP
Catalog No. 501974752
Encompass
Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™). The blunt end reduces the risk of needle-stick injuries. Product is manufactured from non-pyrogenic materials.
Catalog No. 50-197-4752 Supplier STEMCELL Technologies Supplier No. 28120
Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™). The blunt end reduces the risk of needle-stick injuries. Product is manufactured from non-pyrogenic materials.