Learn More
Blunt Forceps
SCI_ED
Catalog No. p-4908183 Shop All Medco Instruments Products
Change view
Click to view available options
Tip Style:
Straight, Serrated
Style:
Straight
Product Type:
Forceps, Dissecting
Catalog No. Tip Style Style Product Type
S17329 Straight, Serrated Straight Forceps, Dissecting
1 options
Catalog No. S17329 Supplier Medco Instruments Supplier No. 82200
Steel construction ensures years of service in every kind of dissecting work.
  • The inside points on these forceps are deeply file-cut to ensure a tight grip
Specifications
Tip Style Straight, Serrated
Length (English) 4.5 in.
Material Stainless Steel
Length (Metric) 114 mm
Product Type Forceps, Dissecting
Quantity (No. per Case) Each
Style Straight