Learn More
Fine Science Tools Micro Hooks, Tip: 0.3 mm, Stainless Steel, 12 cm
SDP
Catalog No. NC9392041
Encompass
Product: Blunt Hook- 12cm/0.3mm Tip Diameter/ Tip Shape: Blunt Hook/ Tip Diameter: 0.3mm/ Alloy: Stainless Steel12cm/ Feature: Each
Catalog No. NC9392041 Supplier Fine Science Tools Supplier No. 1006212
Product: Blunt Hook- 12cm/0.3mm Tip Diameter/ Tip Shape: Blunt Hook/ Tip Diameter: 0.3mm/ Alloy: Stainless Steel12cm/ Feature: Each