Learn More
SAI Infusion Technologies 27 Gauge, 0.5" Blunt Needles with Injection Cap - Sterile, 25/pk
SDP
Catalog No. NC2243849
Encompass
27G0.5" Sterile Blunt Needles with attached Injection Cap. Blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters. Remove the Injection Cap to access the luer directly, or use a Huber needle to pierce the cap without removing it. The cap will prevent the free flow of blood and also reduce the risk of air and pathogens entering the catheter. Sterile, 25/pk. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC2243849 Supplier SAI Infusion Technologies Supplier No. B2750IC
27G0.5" Sterile Blunt Needles with attached Injection Cap. Blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters. Remove the Injection Cap to access the luer directly, or use a Huber needle to pierce the cap without removing it. The cap will prevent the free flow of blood and also reduce the risk of air and pathogens entering the catheter. Sterile, 25/pk. Product is for laboratory animal research only.