Learn More
Cml Supply Blunt Needle 18GAx2 Pink 50pc
Catalog No. 502963916
Encompass_Preferred
18 Gauge 2-in length Blunt Tip Stainless Steel Dispense Needles. Fits all standard Luer Lock or Luer Slip Syringes. Compatible with EFD, Nordson, Techon, PVA dispensing Systems. UV safe plastic hub with luer taper. Non-sterile, but may be sterilized by autoclave. For Industrial and laboratory use only.
Catalog No. 50-296-3916 Supplier Cml Supply Supplier No. 90118200L
18 Gauge 2-in length Blunt Tip Stainless Steel Dispense Needles. Fits all standard Luer Lock or Luer Slip Syringes. Compatible with EFD, Nordson, Techon, PVA dispensing Systems. UV safe plastic hub with luer taper. Non-sterile, but may be sterilized by autoclave. For Industrial and laboratory use only.