Learn More
SAI Infusion Technologies 20 Gauge, 1.5" Blunt Needles - Non-sterile, 1000/pk
SDP
Catalog No. NC0940361
Encompass_Preferred
20G1.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 1000. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC0940361 Supplier SAI Infusion Technologies Supplier No. B20150 1000 BULK
20G1.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 1000. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.