Learn More
Braintree Scientific Blunt Tip Needle non-sterile,18ga - 1"""" w/standard hub (doz)
SDP
Catalog No. 501955352
Encompass
Stainless steel, blunt tip, hypodermic needle with standard luer hub. For use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, blood sampling and catheter flushing. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5352 Supplier Braintree Scientific Supplier No. NB18G
 Restricted Item
Add to Cart
Add to Cart
Stainless steel, blunt tip, hypodermic needle with standard luer hub. For use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, blood sampling and catheter flushing. **Sold for research purposes only and not for human use**