Learn More
Braintree Scientific Blunt Tip Needle non-sterile,20ga -1"""" w/standard hub (dozen
SDP
Catalog No. 501955354
Encompass
Stainless steel, blunt tip, hypodermic needle with standard luer hub. For use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, blood sampling and catheter flushing. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5354 Supplier Braintree Scientific Supplier No. NB20G
Stainless steel, blunt tip, hypodermic needle with standard luer hub. For use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, blood sampling and catheter flushing. **Sold for research purposes only and not for human use**