Learn More
Scientific Commodities Inc Blunt Tip Laboratory Pipetting Needles, 3/pk
Catalog No. NC1380302
Encompass_Preferred
All needles are made of 304 stainless steel and fitted with American standard short female luer hubs of nickel-plated brass that meet ISO 594-2. They are also compliant with current EU RoHS directive 2011/65/EU. The short hub reduces the dead volume inherent in all American standard luer hubs. Needles are supplied with a 90-degree blunt tip.
Catalog No. NC1380302 Supplier Scientific Commodities Inc Supplier No. BB829218
All needles are made of 304 stainless steel and fitted with American standard short female luer hubs of nickel-plated brass that meet ISO 594-2. They are also compliant with current EU RoHS directive 2011/65/EU. The short hub reduces the dead volume inherent in all American standard luer hubs. Needles are supplied with a 90-degree blunt tip.