Learn More
Alkali Scientific Bluo Gal [5 Bromo 3 indolyl B D galactopyranoside], 1 G
SDP
Catalog No. 5000660694
Encompass_Preferred
Bluo Gal (5-Bromo 3-Indolyl β-D-Galactopyranoside) is a chromogenic substrate used to detect β-galactosidase activity in molecular biology applications. The 1g bottle provides sufficient quantity for multiple experiments, making it ideal for laboratories performing gene expression analysis or microbial screening. When β-galactosidase cleaves Bluo Gal, a blue product is formed, indicating the presence of active enzyme. Bluo Gal is particularly valuable in cloning experiments, where it helps identify successfully transformed cells that express β-galactosidase. It can also be used in enzyme activity assays to study the functionality of β-galactosidase in various biological systems.
Catalog No. 50-006-60694 Supplier Alkali Scientific Supplier No. BLU002CP
Bluo Gal (5-Bromo 3-Indolyl β-D-Galactopyranoside) is a chromogenic substrate used to detect β-galactosidase activity in molecular biology applications. The 1g bottle provides sufficient quantity for multiple experiments, making it ideal for laboratories performing gene expression analysis or microbial screening. When β-galactosidase cleaves Bluo Gal, a blue product is formed, indicating the presence of active enzyme. Bluo Gal is particularly valuable in cloning experiments, where it helps identify successfully transformed cells that express β-galactosidase. It can also be used in enzyme activity assays to study the functionality of β-galactosidase in various biological systems.