Learn More
Alkali Scientific Bluo Gal [5 Bromo 3 indolyl B D galactopyranoside], 100 Mg
SDP
Catalog No. 5000660286
Encompass_Preferred
Bluo Gal (5-Bromo 3-Indolyl β-D-Galactopyranoside) is a chromogenic substrate used for detecting β-galactosidase activity in molecular biology applications. This compound produces a blue color when cleaved by β-galactosidase, making it useful for screening recombinant cells, gene expression studies, and enzyme assays. The 100mg quantity is ideal for small-scale experiments or for use in educational settings. Bluo Gal is particularly useful in microbiology and molecular biology labs where monitoring gene expression and enzymatic activity is necessary. This reagent allows for quick visual confirmation of enzyme activity, facilitating the analysis of gene regulation and transformation events in cells.
Catalog No. 50-006-60286 Supplier Alkali Scientific Supplier No. BLU001CP
Bluo Gal (5-Bromo 3-Indolyl β-D-Galactopyranoside) is a chromogenic substrate used for detecting β-galactosidase activity in molecular biology applications. This compound produces a blue color when cleaved by β-galactosidase, making it useful for screening recombinant cells, gene expression studies, and enzyme assays. The 100mg quantity is ideal for small-scale experiments or for use in educational settings. Bluo Gal is particularly useful in microbiology and molecular biology labs where monitoring gene expression and enzymatic activity is necessary. This reagent allows for quick visual confirmation of enzyme activity, facilitating the analysis of gene regulation and transformation events in cells.