Learn More
Alkali Scientific Bluo Gal [5 Bromo 3 indolyl B D galactopyranoside], 5 G
SDP
Catalog No. 5000661672
Encompass_Preferred
Bluo Gal (5-Bromo 3-Indolyl β-D-Galactopyranoside) is a powerful chromogenic substrate used for detecting β-galactosidase activity in molecular biology assays. The 5g bottle is ideal for laboratories performing large-scale experiments or ongoing research. When cleaved by β-galactosidase, Bluo Gal produces a blue color, making it easy to identify and quantify enzyme activity. This reagent is widely used in cloning, gene expression analysis, and bacterial screening, where monitoring β-galactosidase activity is crucial. The 5g size provides an ample supply for continuous use in microbiological and biochemical studies, helping researchers and technicians identify transformed or engineered cells that express β-galactosidase.
Catalog No. 50-006-61672 Supplier Alkali Scientific Supplier No. BLU003CP
Bluo Gal (5-Bromo 3-Indolyl β-D-Galactopyranoside) is a powerful chromogenic substrate used for detecting β-galactosidase activity in molecular biology assays. The 5g bottle is ideal for laboratories performing large-scale experiments or ongoing research. When cleaved by β-galactosidase, Bluo Gal produces a blue color, making it easy to identify and quantify enzyme activity. This reagent is widely used in cloning, gene expression analysis, and bacterial screening, where monitoring β-galactosidase activity is crucial. The 5g size provides an ample supply for continuous use in microbiological and biochemical studies, helping researchers and technicians identify transformed or engineered cells that express β-galactosidase.