Learn More
Research Products International Corp Bluo-Gal [5-Bromo-3-indolyl-B-D-galactopyranoside], 5 Grams
SDP
Catalog No. 501368131
Encompass_Preferred
Chromogenic substrate for B-D-Galactosidase. Produces a deeper blue water insoluble color than X-GAL.
Catalog No. 50-136-8131 Supplier Research Products International Corp Supplier No. B725005.0
Chromogenic substrate for B-D-Galactosidase. Produces a deeper blue water insoluble color than X-GAL.