Learn More
Sigma Aldrich Fine Chemicals Biosciences Bluo Gal reagen for selec500MG
Catalog No. 502903126
Encompass
Bluo-Gal is a chromogenic substrate for beta-galactosidase used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Bluo-Gal is designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac phenotype.
Catalog No. 50-290-3126 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B2904500MG
Bluo-Gal is a chromogenic substrate for beta-galactosidase used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Bluo-Gal is designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac phenotype.