Learn More
Aviva Systems Biology Corp BLVRB Antibody - middle region (ARP56121_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56121_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BLVRB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BLVRB. Alternate names: BVRB, FLR, MGC117413, SDR43U1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61727