Learn More
Sino Biological Recombinant Anti-BLVRB Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13151R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human BLVRB / biliverdin reductase B (rh BLVRB / biliverdin reductase B; Catalog#13151-H07E; P30043; Ala2-Gln206).
Catalog No. 50-224-7919