Learn More
BioVendor, LLC Blyscan Dye Dissoc 2X2X110Ml BioVendor LLC
SDP
Catalog No. 501725942
Encompass
TypeColorimetric assayApplicationsCell culture and/or animal studies, Urine, Synovial fluid, Tissue extractShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationAll components are stable for one year, (from Invoice Date), when stored at 15¿25¿C. Do not store below +4¿C. Once opened the glass vial containing sGAG standard should however be stored at +4¿C.
Catalog No. 50-172-5942 Supplier BioVendor, LLC Supplier No. B1500
TypeColorimetric assayApplicationsCell culture and/or animal studies, Urine, Synovial fluid, Tissue extractShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationAll components are stable for one year, (from Invoice Date), when stored at 15¿25¿C. Do not store below +4¿C. Once opened the glass vial containing sGAG standard should however be stored at +4¿C.