Learn More
BioVendor, LLC Blyscan; Dye Reagent, (3 x 110 ml)
SDP
Catalog No. 501725938
Encompass
Blyscan; Dye Reagent, (3 x 110 ml). Contains 1, 9-dimethyl-methylene blue in an inorganic buffer, which also contains surfactants. Blyscan™ sGAG Assay dye component. Biocolor Item # B1005
Catalog No. 50-172-5938 Supplier BioVendor, LLC Supplier No. B1005
Blyscan; Dye Reagent, (3 x 110 ml). Contains 1, 9-dimethyl-methylene blue in an inorganic buffer, which also contains surfactants. Blyscan™ sGAG Assay dye component. Biocolor Item # B1005