Learn More
BioVendor, LLC Blyscan Sgag 110 Assay Stdsize BioVendor LLC
SDP
Catalog No. 501725937
Encompass
TypeColorimetric assayApplicationsCell culture and/or animal studies, Urine, Synovial fluid, Tissue extractSample Requirements10 ¿ 100 ¿lShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationAll components are stable for one year, (from Invoice Date), when stored at 15¿25¿C. Do not store below +4¿C. Once opened the glass vial containing sGAG standard should however be stored at +4¿C. Calibration Range1 ¿ 5 ¿gLimit of Detection0.25 ¿g
Catalog No. 50-172-5937 Supplier BioVendor, LLC Supplier No. B1000
TypeColorimetric assayApplicationsCell culture and/or animal studies, Urine, Synovial fluid, Tissue extractSample Requirements10 ¿ 100 ¿lShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationAll components are stable for one year, (from Invoice Date), when stored at 15¿25¿C. Do not store below +4¿C. Once opened the glass vial containing sGAG standard should however be stored at +4¿C. Calibration Range1 ¿ 5 ¿gLimit of Detection0.25 ¿g