Learn More
ILEX LIFE SCIENCES LLC BLYSCAN SGAG ISO/CONC REAGENTS
SDP
Catalog No. 502733955
Encompass_Preferred
Biocolor Blyscan Glycosaminoglycan (sGAG) Isolation & Concentration Pack (100 samples). DESCRIPTION: Isolation & Concentration reagents for use with Blyscan Glycosaminoglycan Assay. Blyscan Glycosaminoglycan Assay is a quantitative colorimetric dye-binding method for analysis of sulfated proteoglycans and glycosaminoglycans (sGAG). It can be used to measure total sGAG content and can also to determine the O- and N-sulfated GAG ratio within test samples. The assay uses 1,9-dimethylmethylene blue dye employed under conditions that provide a specific label for the sulfated polysaccharide component of proteoglycans or the protein free sulfated glycosaminoglycan chains. APPLICATIONS: Cell culture derived material, soluble sGAGs from fluids (ex. synovial fluid, urine), fibrous and hyaline cartilages, tissues (ex. arteries, heart, lung, skin, tumors). Soluble sGAGs may be assayed directly, solid materials will require a preliminary papain extraction.
Catalog No. 50-273-3955 Supplier ILEX LIFE SCIENCES LLC Supplier No. B1015
Biocolor Blyscan Glycosaminoglycan (sGAG) Isolation & Concentration Pack (100 samples). DESCRIPTION: Isolation & Concentration reagents for use with Blyscan Glycosaminoglycan Assay. Blyscan Glycosaminoglycan Assay is a quantitative colorimetric dye-binding method for analysis of sulfated proteoglycans and glycosaminoglycans (sGAG). It can be used to measure total sGAG content and can also to determine the O- and N-sulfated GAG ratio within test samples. The assay uses 1,9-dimethylmethylene blue dye employed under conditions that provide a specific label for the sulfated polysaccharide component of proteoglycans or the protein free sulfated glycosaminoglycan chains. APPLICATIONS: Cell culture derived material, soluble sGAGs from fluids (ex. synovial fluid, urine), fibrous and hyaline cartilages, tissues (ex. arteries, heart, lung, skin, tumors). Soluble sGAGs may be assayed directly, solid materials will require a preliminary papain extraction.