Learn More
BioVendor, LLC Blyscan; sGAG Reference Standard (100 ug/ml; 3 x 5 ml).
SDP
Catalog No. 501725940
Encompass
Blyscan; sGAG Reference Standard (100 ug/ml; 3 x 5 ml). Sterile solution of bovine tracheal chondroitin 4-sulfate. Concentration: 100 g/ml. Blyscan™ sGAG (Glycosaminoglycan) Assay reference standard component. Biocolor Item # B1010
Catalog No. 50-172-5940 Supplier BioVendor, LLC Supplier No. B1010
Blyscan; sGAG Reference Standard (100 ug/ml; 3 x 5 ml). Sterile solution of bovine tracheal chondroitin 4-sulfate. Concentration: 100 g/ml. Blyscan™ sGAG (Glycosaminoglycan) Assay reference standard component. Biocolor Item # B1010