Learn More
AdipoGen BLZ-945,Chemical. CAS: 953769-46-5
SDP
Catalog No. 501017050
Encompass_Preferred
BLZ-945, AdipoGen Life Sciences. Chemical. CAS: 953769-46-5. Formula: C20H22N4O3S. MW: 398.5. BLZ945 is an orally active, potent and selective colony stimulating factor 1 receptor (CSF-1R) inhibitor, which inhibits CSF-1R activity with an IC(50) of 1nM and is more than 1000-fold selective against its closest receptor tyrosine kinase homologs c-KIT and Platelet-derived Growth Factor Receptor beta (PDGFRbeta ) as well as more than 200 additional kinases, confirming the selectivity of the compound.
Catalog No. 50-101-7050 Supplier AdipoGen Supplier No. SYN1197M001
BLZ-945, AdipoGen Life Sciences. Chemical. CAS: 953769-46-5. Formula: C20H22N4O3S. MW: 398.5. BLZ945 is an orally active, potent and selective colony stimulating factor 1 receptor (CSF-1R) inhibitor, which inhibits CSF-1R activity with an IC(50) of 1nM and is more than 1000-fold selective against its closest receptor tyrosine kinase homologs c-KIT and Platelet-derived Growth Factor Receptor beta (PDGFRbeta ) as well as more than 200 additional kinases, confirming the selectivity of the compound.