Learn More
AdipoGen BLZ-945 (5 mg)
SDP
Supplier:  AdipoGen AGCR13609M005
Encompass_Preferred
Chemical. CAS: 953769-46-5. Formula: C20H22N4O3S. MW: 398.5. Anticancer compound. Orally active, potent and selective colony stimulating factor 1 receptor (CSF-1R; c-Fms) inhibitor. More than 1000-fold selective against its closest receptor tyrosine kinase homologs c-KIT and platelet-derived growth factor receptor beta (PDGFRbeta). Selective against more than 200 additional kinases.
Catalog No. 50-205-3979