Learn More
Medchemexpress LLC HY-12768 50mg Medchemexpress, BLZ945 CAS:953769-46-5 Purity:>98%
SDP
Catalog No. 502028913
Encompass_Preferred
Medchemexpress, HY-12768 50mg BLZ945 CAS:953769-46-5 BLZ945 is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8913 Supplier Medchemexpress LLC Supplier No. HY1276850MG
Medchemexpress, HY-12768 50mg BLZ945 CAS:953769-46-5 BLZ945 is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.