Learn More
Medchemexpress LLC HY-12768 100mg Medchemexpress, BLZ945 CAS:953769-46-5 Purity:98%
SDP
Catalog No. 501637824
Encompass_Preferred
Medchemexpress,HY-12768 100mg BLZ945 CAS:953769-46-5 Formula:C20H22N4O3S BLZ945 is a potent, selective and brain-penetrant CSF-1R inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7824 Supplier Medchemexpress LLC Supplier No. HY12768100MG
Medchemexpress,HY-12768 100mg BLZ945 CAS:953769-46-5 Formula:C20H22N4O3S BLZ945 is a potent, selective and brain-penetrant CSF-1R inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.