Learn More
Aviva Systems Biology Corp BLZF1 Antibody - N-terminal region (ARP38375_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38375_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BLZF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human BLZF1. Alternate names: JEM1, JEM-1, JEM-1s, GOLGIN-45ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56627